Assay Details
Gene Symbol: MYCBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038565.2 | NP_001033654.1 | 3 - 4 | 142 | 97 | |
GenBank mRNA | BC109848.1 | - | 3 - 4 | 143 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
