Assay Details
Gene Symbol: GLRX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040523.2 | NP_001035613.1 | 3 - 4 | 435 | 112 | |
GenBank mRNA | BC109742.1 | - | 3 - 4 | 436 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
