Assay Details
Gene Symbol: RPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040581.2 | NP_001035671.1 | 233 | 72 | ||
GenBank mRNA | BC108180.1 | - | 234 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
