Assay Details
Gene Symbol: EEF1E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040599.1 | NP_001035689.1 | 2 - 3 | 297 | 146 | |
GenBank mRNA | BC103259.1 | - | 2 - 3 | 297 | 146 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
