Assay Details
Gene Symbol: RPL11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075581.2 | NP_001069049.1 | 1 | 283 | 95 | |
GenBank mRNA | BC102524.1 | - | 1 | 285 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
