Assay Details
Gene Symbol: ROPN1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075717.2 | NP_001069185.1 | 1 - 2 | 352 | 76 | |
GenBank mRNA | BC102601.1 | - | 1 - 2 | 354 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC102601.1, NM_001075717.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
