Assay Details
Gene Symbol: RPL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076998.2 | NP_001070466.1 | 6 - 7 | 426 | 79 | |
GenBank mRNA | BC103039.1 | - | 6 - 7 | 427 | 79 | |
BC105558.1 | - | 4 - 5 | 1212 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
