Assay Details
Gene Symbol: SSR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077044.2 | NP_001070512.1 | 1 - 2 | 175 | 129 | |
GenBank mRNA | BC103053.1 | - | 1 - 2 | 176 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
