Assay Details
Gene Symbol: SLC9A3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077852.2 | NP_001071320.1 | 3 - 4 | 1010 | 99 | |
GenBank mRNA | BC102807.1 | - | 3 - 4 | 1011 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
