Assay Details
Gene Symbol: SLC9A3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077852.2 | NP_001071320.1 | 5 - 6 | 1146 | 70 | |
GenBank mRNA | BC102807.1 | - | 5 - 6 | 1147 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
