Assay Details
Gene Symbol: DCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078023.2 | NP_001071491.1 | 1 - 2 | 185 | 97 | |
GenBank mRNA | BC103167.1 | - | 1 - 2 | 186 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
