Assay Details
Gene Symbol: GTF2A1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080229.1 | NP_001073698.1 | 2 - 3 | 207 | 79 | |
GenBank mRNA | BC105172.1 | - | 2 - 3 | 207 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
