Assay Details
Gene Symbol: RXRB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083640.1 | NP_001077109.1 | 4 - 5 | 813 | 93 | |
GenBank mRNA | BC146260.1 | - | 4 - 5 | 992 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
