Assay Details
Gene Symbol: PAEP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173929.3 | NP_776354.2 | 2 - 3 | 271 | 69 | |
GenBank mRNA | BC108213.1 | - | 2 - 3 | 271 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
