Assay Details
Gene Symbol: CXADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174298.4 | NP_776723.1 | 1 - 2 | 154 | 93 | |
GenBank mRNA | AY033651.1 | - | 1 - 2 | 45 | 93 | |
BC102845.1 | - | 1 - 2 | 155 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
