Assay Details
Gene Symbol: CXADR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174298.4 | NP_776723.1 | 4 - 5 | 675 | 96 | |
GenBank mRNA | AY033651.1 | - | 4 - 5 | 566 | 96 | |
BC102845.1 | - | 4 - 5 | 676 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
