Assay Details
Gene Symbol: NEDD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174764.3 | NP_777189.1 | 2 - 3 | 134 | 71 | |
GenBank mRNA | AF227256.1 | - | 2 - 3 | 99 | 71 | |
BC102306.1 | - | 2 - 3 | 134 | 71 | ||
DQ347574.1 | - | 1 - 2 | 43 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
