Assay Details
Gene Symbol: NEDD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174764.3 | NP_777189.1 | 3 - 4 | 213 | 87 | |
GenBank mRNA | AF227256.1 | - | 3 - 4 | 178 | 87 | |
BC102306.1 | - | 3 - 4 | 213 | 87 | ||
DQ347598.1 | - | 1 - 2 | 35 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
