Assay Details
Gene Symbol: MMACHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015588.3 | NP_001015588.2 | 3 - 4 | 558 | 70 | |
GenBank mRNA | BC110137.1 | - | 3 - 4 | 559 | 70 | |
BT020992.1 | - | 3 - 4 | 435 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
