Assay Details
Gene Symbol: RASSF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024549.2 | NP_001019720.1 | 3 - 4 | 1178 | 113 | |
GenBank mRNA | BC111208.1 | - | 3 - 4 | 1178 | 113 | |
BT021890.1 | - | 2 - 3 | 1070 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
