Assay Details
Gene Symbol: BUD23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034457.2 | NP_001029629.2 | 5 - 6 | 385 | 97 | |
GenBank mRNA | BC109889.1 | - | 5 - 6 | 385 | 97 | |
BT021557.1 | - | 5 - 6 | 410 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
