Assay Details
Gene Symbol: GRINA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037593.1 | NP_001032682.1 | 6 - 7 | 1053 | 102 | |
GenBank mRNA | BC109761.1 | - | 6 - 7 | 1053 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
