Assay Details
Gene Symbol: CD86 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038017.2 | NP_001033106.1 | 5 - 6 | 931 | 102 | |
GenBank mRNA | AY533858.1 | - | 1 - 2 | 130 | 102 | |
BT025428.1 | - | 5 - 6 | 987 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
