Assay Details
Gene Symbol: AARSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038073.1 | NP_001033162.1 | 5 - 6 | 624 | 69 | |
GenBank mRNA | BC109537.1 | - | 5 - 6 | 624 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
