Assay Details
Gene Symbol: DTD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038104.2 | NP_001033193.1 | 3 - 4 | 457 | 118 | |
GenBank mRNA | BC111244.1 | - | 3 - 4 | 458 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
