Assay Details
Gene Symbol: METTL26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038106.2 | NP_001033195.1 | 2 - 3 | 539 | 66 | |
GenBank mRNA | BC109870.1 | - | 2 - 3 | 541 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
