Assay Details
Gene Symbol: CNN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038179.1 | NP_001033268.1 | 1 | 291 | 107 | |
GenBank mRNA | BC109697.1 | - | 1 | 291 | 107 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109697.1, NM_001038179.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
