Assay Details
Gene Symbol: DMRTC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038182.2 | NP_001033271.1 | 5 - 6 | 698 | 115 | |
GenBank mRNA | BC109620.1 | - | 5 - 6 | 699 | 115 | |
EF534775.1 | - | 5 - 6 | 689 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
