Assay Details
Gene Symbol: SPRYD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038206.2 | NP_001033295.1 | 1 - 2 | 333 | 104 | |
GenBank mRNA | BC111224.1 | - | 1 - 2 | 334 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
