Assay Details
Gene Symbol: PMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038212.2 | NP_001033301.1 | 2 - 3 | 295 | 98 | |
GenBank mRNA | BC111342.1 | - | 2 - 3 | 296 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
