Assay Details
Gene Symbol: TRAPPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191204.1 | NP_001178133.1 | 3 - 4 | 799 | 59 | |
GenBank mRNA | BC109939.1 | - | 3 - 4 | 477 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
