Assay Details
Gene Symbol: VBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038516.2 | NP_001033605.1 | 1 - 2 | 143 | 115 | |
GenBank mRNA | BC109526.1 | - | 1 - 2 | 144 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
