Assay Details
Gene Symbol: VBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038516.2 | NP_001033605.1 | 5 - 6 | 564 | 156 | |
GenBank mRNA | BC109526.1 | - | 5 - 6 | 565 | 156 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
