Assay Details
Gene Symbol: ADPRH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038554.1 | NP_001033643.1 | 2 - 3 | 426 | 118 | |
GenBank mRNA | BC109957.1 | - | 2 - 3 | 426 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
