Assay Details
Gene Symbol: SMU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038573.1 | NP_001033662.1 | 5 - 6 | 656 | 102 | |
GenBank mRNA | BC109712.1 | - | 5 - 6 | 656 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
