Assay Details
Gene Symbol: ETFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038582.1 | NP_001033671.1 | 4 - 5 | 466 | 78 | |
GenBank mRNA | BC109603.1 | - | 4 - 5 | 466 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
