Assay Details
Gene Symbol: AICDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038682.3 | NP_001033771.2 | 2 - 3 | 232 | 81 | |
GenBank mRNA | DQ303466.1 | - | 1 - 2 | 159 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
