Assay Details
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038686.2 | NP_001033775.1 | 5 - 6 | 409 | 114 | |
GenBank mRNA | BC110230.1 | - | 5 - 6 | 410 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
