Assay Details
Gene Symbol: PRSS37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040569.2 | NP_001035659.1 | 5 - 6 | 671 | 129 | |
GenBank mRNA | BC109928.1 | - | 5 - 6 | 672 | 129 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
