Assay Details
Gene Symbol: DYDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040571.2 | NP_001035661.1 | 1 - 2 | 75 | 109 | |
GenBank mRNA | BC109581.1 | - | 1 - 2 | 76 | 109 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC109581.1, NM_001040571.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
