Assay Details
Gene Symbol: DYDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040571.2 | NP_001035661.1 | 4 - 5 | 488 | 113 | |
GenBank mRNA | BC109581.1 | - | 4 - 5 | 489 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
