Assay Details
Gene Symbol: CPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045889.2 | NP_001039354.1 | 1 - 2 | 267 | 93 | |
GenBank mRNA | BC105423.1 | - | 1 - 2 | 268 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
