Assay Details
Gene Symbol: TRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045896.2 | NP_001039361.1 | 3 - 4 | 509 | 84 | |
GenBank mRNA | BC112743.1 | - | 3 - 4 | 510 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
