Assay Details
Gene Symbol: GADD45G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045901.1 | NP_001039366.1 | 1 - 2 | 160 | 68 | |
GenBank mRNA | BC112476.1 | - | 1 - 2 | 160 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
