Assay Details
Gene Symbol: APOA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045916.2 | NP_001039381.1 | 2 - 3 | 109 | 62 | |
GenBank mRNA | BC111607.1 | - | 2 - 3 | 111 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
