Assay Details
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045962.1 | NP_001039427.1 | 5 - 6 | 654 | 120 | |
GenBank mRNA | BC110260.1 | - | 5 - 6 | 654 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
