Assay Details
Gene Symbol: ACYP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045982.2 | NP_001039447.1 | 1 - 2 | 89 | 105 | |
GenBank mRNA | BC114901.1 | - | 1 - 2 | 90 | 105 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC114901.1, NM_001045982.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
