Assay Details
Gene Symbol: SF3B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046017.1 | NP_001039482.1 | 1 - 2 | 255 | 117 | |
GenBank mRNA | BC112548.1 | - | 1 - 2 | 255 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC112548.1, NM_001046017.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
