Assay Details
Gene Symbol: ASPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046033.2 | NP_001039498.1 | 6 - 7 | 840 | 131 | |
GenBank mRNA | BC112799.1 | - | 6 - 7 | 841 | 131 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
