Assay Details
Gene Symbol: CYRIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046047.2 | NP_001039512.1 | 7 - 8 | 618 | 125 | |
GenBank mRNA | BC105327.1 | - | 7 - 8 | 620 | 125 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
